n-Heneicosane 98%

Supplier: Thermo Scientific Chemicals

35277-0500 35277-0100 35277-0010
TS35277-0010EA 32.04 USD
TS35277-0010 TS35277-0100 TS35277-0500
n-Heneicosane 98%
n-Heneicosane
Formula: H₃C(CH₂)₁₉CH₃
MW: 296.58 g/mol
Boiling Pt: 100 °C (2 mmHg)
Melting Pt: 39…43 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009346
CAS Number: 629-94-7
EINECS: 211-118-9

Order Now

Specification Test Results

Appearance White Crystalline chunks or crystals or flakes
Infrared spectrum Conforms
Melting point 39 to 43 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR