n-Eicosane 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Eicosane

20437-1000 20437-5000 20437-0020
TS20437-1000EA 69.47 USD
TS20437-1000 TS20437-0020 TS20437-5000
n-Eicosane 99%
n-Eicosane
Formula: H₃C(CH₂)₁₈CH₃
MW: 282.55 g/mol
Boiling Pt: 220 °C (30 mmHg)
Melting Pt: 36…38 °C
Density: 0.7886 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00009344
CAS Number: 112-95-8
EINECS: 204-018-1

Order Now

Specification Test Results

Appearance Transparent white Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 36 to 38 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR