1-Bromooctadecane 96%

Supplier: Thermo Scientific Chemicals

19002-0025 19002-5000 19002-1000
TS19002-1000EA 37 USD
TS19002-1000 TS19002-0025 TS19002-5000
1-Bromooctadecane 96%
1-Bromooctadecane
Formula: BrH₂C(CH₂)₁₆CH₃
MW: 333.4 g/mol
Boiling Pt: 214…216 °C (12 mmHg)
Melting Pt: >20 °C
Density: 0.97 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000231
CAS Number: 112-89-0
EINECS: 204-013-4

Order Now

Specification Test Results

Appearance White to yellow Fused solid or liquid
Infrared spectrum Conforms
Melting point 25 to 31 °C
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR