Linktech 40AC and 40PP Quick-Disconnect Fitting Insert, BSPT(M) without Valve
Supplier: VWR International
40AC-PB1-04BSPT
MFLX41304-60EA 48.5 USD
MFLX41304-60
Linktech 40AC and 40PP Quick-Disconnect Fitting Insert, BSPT(M) without Valve
Fittings
Easy to disconnect from 1/4" flow line with one hand.
  • Withstands pressures from vacuum to 120 psi (8.3 bar)
  • Interchangeable with other similar industry thumb-latch fittings
Just press the metal thumb-latch to connect and disconnect fitting halves. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized.
Mix and match valved and straight-through (nonvalved) fittings to suit your flow path needs—simply order a body (female half) and an insert (male half). Valved fittings automatically seal the flow path when disconnected.