LinkTech 60PP and 60PS Quick-Disconnect Fitting Body, Hosebarb with Valve
Supplier: VWR International
60PPV-SE2-08
MFLX41303-10EA 119 USD
MFLX41303-10
LinkTech 60PP and 60PS Quick-Disconnect Fitting Body, Hosebarb with Valve
Fittings
Disconnect fittings quickly with large plastic thumb-latch.
  • Audible click lets you know the fittings are secure
  • Buna N O-ring seal eliminates leaks
  • Interchangeable with other similar industry thumb-latch fittings
Easily connect and disconnect from your 3/8” flow line with the 60PP and PS quick-disconnect fittings. Just press the robust plastic thumb-latch to quickly disconnect fitting halves. Mix and match valved and straight-through (nonvalved) fittings to suit your flowpath needs—simply order a body (female half) and an insert (male half).
Choose between polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized or polysulfone (PSF) for a wide operating temperature range and greater durability. Valved fittings automatically seal the flow path when disconnected.