Economical Submersible Pumps, Mild Acid Specialty
Supplier: Avantor Fluid Handling
502303 501020 518020
MFLX75501-51EA 158 USD
MFLX75501-51 MFLX75501-53 MFLX75501-57
Economical Submersible Pumps, Mild Acid Specialty
Pumps Submersible Pumps and Sump Pumps
Versatile – operate pumps in either a vertical or horizontal position.
  • Compact and economical
  • Screened inlet helps prevent clogging
Mild acid specialty pumps for use with solutions compatible with pump materials.
Delivery information: Includes 6 ft (1.8 m) cord; 115 VAC models include grounded plug.