Economical Submersible Pumps, High-Flow Utility
Supplier: Avantor Fluid Handling
505025 505202
MFLX75500-05EA 335 USD
MFLX75500-05 MFLX75500-00
Economical Submersible Pumps, High-Flow Utility
Pumps Submersible Pumps and Sump Pumps
Versatile – operate pumps in either a vertical or horizontal position.
  • Compact and economical
  • Screened inlet helps prevent clogging
High-flow submersible utility pumps for use for water transfer with garden hose adpater or recirculation, water displays, air conditioners, and machine tool coolants. Removable inlet screen allows for water removal 1/8" from floor. Oil-filled for longer motor life. UL listed, CSA certified.
Note: Pumps must be fully submersed when operating and not for use with salt water or flammable solutions.
Delivery information: 115 VAC models include grounded plug.