Kobold® Float Type Flow Switches, Brass
Supplier: Kobold
S-3108
MFLX32481-45EA 309 USD
MFLX32481-45
Kobold® Float Type Flow Switches, Brass
Flow Switches
Small, compact design with low weight.
  • Constructed of durable brass
The device works on the well known float principle but without the use of the open conical measuring tube. Instead, the instruments use a cylindrical control tube with a conical groove along its axis. The flow switch can be used for continuous control of fluids and gases to ensure that excess or low flow cannot result in damage to the equipment. Applications can be found wherever it is necessary to control flow reliably.
FEATURES
- ± 4% Full scale accuracy
- Small, compact design, low weight
- Direct reading scales for water and air
- Large and small measuring ranges from 28:1 to 2.6:1 turndown
- Vertical connections for easy installation
- All media contacted parts available in brass or stainless steel construction