Flowline Buoyancy Vertical-Mount Level Switch
Supplier: Flowline
AV16-4243-A AV13-4213-A
MFLX43305-10EA 599 USD
MFLX43305-10 MFLX68873-76
Flowline Buoyancy Vertical-Mount Level Switch
Level Sensors
Don’t let your tanks overflow.
  • Optional strobe alarm alerts you to high level conditions
  • Fail-safe relay control of pumps and valves with 0 to 60 second time delay
  • Buoyancy switches are ideal for clean water and noncoating chemicals
  • NEMA 4X rated enclosure with 300° conduit swivel
  • Select length for custom set points
Switch-Pak™, a general-purpose switch package, provides a complete solution for high level spill prevention with a fixed level switch point and 10 A compact relay controller for pump or valve actuation. These products are typically selected for corrosive bulk storage, day tank and waste sump applications.
These switches are available in 6" to 10-ft lengths, specified in 1/2" increments. Must specify length "A" when ordering.