CINC V2 Centrifugal Multipurpose Separator/Extractors
Supplier: CINC
V-2CP 220V V-2CP W/XP MOTER-110V V-2CP WITH 110V OPTION V-2CP W/XP MOTOR
MFLX17315-06EA 9679 USD
MFLX17315-06 MFLX17315-04 MFLX17315-02 MFLX17315-00
CINC V2 Centrifugal Multipurpose Separator/Extractors
Pumps Centrifugal Separators
Mix, react, and/or separate liquid-liquid mixtures in a single device.
  • Highly flexible design operates effectively under changing inlet concentrations, variable flow rates, and flow disruptions
  • Heavy-duty SS componentry can be quickly cleaned and inspected (suitable for CIP and autoclavable)
  • Interchangeable weirs allow for broad applications in research, pilot plant, or production operations
Users find this design to be superior to most disk-stack style centrifuges, gravity-based decanters, coalescer and DAF type separators, mixer/settler systems, and extraction columns. It is suitable for separation of liquids with specific gravity differences of 0.05 or better and can process immiscible droplet sizes that are up to 10 ìm. Users may interchange weirs to accommodate for liquids of varying specific gravity and viscosity. A second inlet port allows the introduction (down to 20 ml/min) of mixing additives to the liquid for extraction, reaction and washing.
Delivery information: Includes centrifugal chamber, 1⁄8-hp electric motor with variable speed controller, high- and low-mix bottom plates plus ten weirs (ranging 0.825" to 1.050" dia), and a CD-ROM including weir selection software, dynamic-image instruction package, and maintenance.