316 Stainless Steel Investment Cast Close-Coupled Standard Centrifugal Pumps
Supplier: SHERWOOD/HYPRO DIVISION
COMSV33 COMSV11 COMSV55 COMSV443
MFLX73031-40EA 1490.25 USD
MFLX73031-40 MFLX73031-20 MFLX73031-00 MFLX73031-35
316 Stainless Steel Investment Cast Close-Coupled Standard Centrifugal Pumps
Pumps Centrifugal Pumps
Open impellers handle clean fluids and fluids containing suspended solids.
  • Heavy-duty, stainless steel design—ideal for aggressive chemicals and high purity solutions
  • Rotate head in 90° increments for multiple mounting configurations
  • Viton® mechanical seal offers superior chemical resistance and accepts high inlet pressures
These close-coupled centrifugal pumps are constructed with a 316 stainless steel (SS) pump head, impeller, and shaft, along with Viton seals with carbon and ceramic faces. The open impeller design can handle clean fluids and fluids containing suspended solids.