Parker Hannifin Poppet Check Valves, PFA
Supplier: Parker Hannifin
CV-1-1144 CV-1-1188
MFLX06373-57EA 352.13 USD
MFLX06373-57 MFLX06373-59
Parker Hannifin Poppet Check Valves, PFA
Valves Check Valves
Install in any position.
  • Constructed of chemically inert PFA with Viton O-rings
Valves have body made of PFA and O-ring seals made of Viton®. Max pressure is 100 psi; temperature range is 0 to 300 °F (–18 to 149 °C). Install in any position. Note: Valves require 5 psi back pressure to achieve a proper seal.