Wash-down Duty NEMA Type C-Face Rigid Base Motors
56T17V5336/ N567 56C17W5331/ N557
MFLX71008-25EA 724.75 USD
MFLX71008-25 MFLX71009-15
Wash-down Duty NEMA Type C-Face Rigid Base Motors
Pumps Motors
Ideal for high humidity, wet environment, or high-pressure wash-down applications.
  • 303 stainless steel fan guard
  • Frame, brackets, and conduit box are coated with USDA-approved white epoxy paint
Single-phase motors have a capacitor. Three-phase motors are rated for inverter duty.