304 Stainless Steel NEMA Type C-Face Rigid-Base Motors
Supplier: VWR International
191475 N411
MFLX71012-25EA 1088 USD
MFLX71012-25 MFLX71015-25
304 Stainless Steel NEMA Type C-Face Rigid-Base Motors
Pumps Motors
Use in pharmaceutical and food processing facilities.
  • Washdown construction capable of inverter duty (range: 30 to 70Hz)
  • Antibacterial, antifungal, and antimoisture treatments
  • Suitable for chemical processes involving nitric acid and caustic lye
Motors operate at 50/60 Hz, if 50 Hz then 190/380 VAC rated at next lower horsepower.