Parker High-Pressure Metal Two-way Ball Valves (Straight)
Supplier: Parker
4A-MB4LPFA-BP 2F-MB4LPFA-BP 8F-MB8LPFA-SSP 4F-MB6LPFA-SSP 6A-MB6LPFA-SSP 2A-MB2LPFA-BP 4F-MB6LPFA-BP 4M-MB6LPFA-SSP 2A-MB2LPFA-SSP 4A-MB4LPFA-SSP 2F-MB4LPFA-SSP
MFLX30527-70EA 153 USD
MFLX30527-70 MFLX30527-36 MFLX30527-38 MFLX30527-66 MFLX30527-44 MFLX30527-32 MFLX30527-68 MFLX30527-34 MFLX30527-40 MFLX30527-64 MFLX30527-30
Parker High-Pressure Metal Two-way Ball Valves (Straight)
Valves Ball Valves
Don’t worry about leaks.
  • Constructed of metal parts for durable applications
Ball valves are designed primarily for on/off service. The two-way valve model has a single inlet port and a single outlet port for straight through operation. The handle of the valve can be turned to change the flow pattern or on/off service of the valve. Choose from stainless steel/PFA or brass/PFA valve options.