Asco Solenoid-Operated 2- and 3-Way Pinch Valves
Supplier: Asco
SCH284A013- 24VDC SCH384A005- 12VDC SCH284B007- 12VDC SCH284B007- 24VDC SCH384A005-24VDC SCH284B015- 12VDC SCH284B015- 24VDC SCH284A013- 12VDC SCH284A005- 24VDC SCH284A005- 12VDC
MFLX01340-61EA 164.61 USD
MFLX01340-61 MFLX01340-60 MFLX01340-57 MFLX01340-56 MFLX01340-67 MFLX01340-66 MFLX01340-55 MFLX01340-54 MFLX01340-63 MFLX01340-62
Asco Solenoid-Operated 2- and 3-Way Pinch Valves
Valves Solenoid Valves
Simple tubing replacement.
  • No contamination of valve
These pinch valves are excellent for isolation-type applications as the only wetted component is the tubing. They feature removable coils with DIN-style electrical connectors and a panel mount bracket is included. Ideal for precise fluid handling applications, these valves have a 20 msec response time.