Positive Displacement Bypass Diaphragm Pumps, Moderate Flow
Supplier: SHURFLO LLC
8020-503-250 8000-543-250
MFLX75420-00EA 261.81 USD
MFLX75420-00 MFLX75420-07
Positive Displacement Bypass Diaphragm Pumps, Moderate Flow
Pumps Positive Displacement Pumps
Ideal for spray nozzles and transfer applications where smooth, consistent flow is required.
  • Constructed of Polypropylene, Santoprene®, and Viton®
Pumps accommodate any size spray orifice. Maintain a near-constant pressure across pumping range for any size nozzle used. A flexible diaphragm design allows pump to run dry without damage.