Hayward® True Union Ball Check Valves with Socket and NPT(F) Threaded Connectors
Supplier: Hayward
TC10100STE TC10200STE TC10075STE TC10150STE TC10050STE 161561101
MFLX01341-10EA 214.04 USD
MFLX01341-10 MFLX01368-10 MFLX01341-00 MFLX01341-02 MFLX01341-04 MFLX01341-08
Hayward® True Union Ball Check Valves with Socket and NPT(F) Threaded Connectors
Valves Check Valves
Unique square-cut elastomer seat provides seal at very low back pressures.
  • Constructed of a PVC body and EPDM seal
Quickly and easily install either vertically or horizontally. These valves are assembled with water soluble, silicone-free lubricant and constructed of a one-piece PVC body.