Hayward Single-Length Bag Filter Housing
Supplier: Hayward
FLV4P2S20T4VS
MFLX29990-04EA 2214.45 USD
MFLX29990-04
Hayward Single-Length Bag Filter Housing
Filters Bag Filter Systems
All-plastic construction—no need to worry about rust and corrosion!
  • Feature hand-removable cover and built-in basket/bag handles—simply lift out the basket and replace the filter bag
  • External cover threads are not in contact with the process media—eliminates the need for cleaning
  • Cover features buttress style threads for extra strength
  • Each filter includes an integral mounting flange and Viton(R) seal; order bags separately.
This Hayward single-length bag filter housing is ideal for water, chemical, pharmaceutical, food and other general fluid filtration applications. Maximum flow rate for double-length size is 100 GPM.