Check and Foot Valves with Hose Barb Connections
Supplier: Bel-Art Products, a part of SP
197160000 19713-0001 197170000 197150000
MFLX06308-10EA 19.89 USD
MFLX06308-10 MFLX06304-30 MFLX06304-20 MFLX06304-10
Check and Foot Valves with Hose Barb Connections
Valves Check Valves
Ideal for low-pressure applications.
  • Easy control of low-pressure applications
Choose from economical LDPE with neoprene disc or autoclavable Polypropylene (PP) with neoprene discs.