FMI® Low-Flow Compact Metering Pumps
Supplier: FMI
RHSY1CKC RHSY0CKC2 RHSY0CKC RHSY1CKC2
MFLX07115-10EA 1136.39 USD
MFLX07115-10 MFLX07115-25 MFLX07115-12 MFLX07115-27
FMI® Low-Flow Compact Metering Pumps
Pumps Piston Pumps
Compact design RH pump with synchronous motor assembly.
  • Drift-free performance independent of load variations or fluctuations in line voltage
  • Micrometer-like fine adjustment using an easy grip flow control ring graduated in 450 divisions
  • Choice of 150, 300, and 600 RPM through a simple and safe belt arrangement change
Forward-Off-Reverse switch for instant flow direction control