Economical Submersible Pumps, Low-Flow Centrifugal
Supplier: Avantor Fluid Handling
527003 527016 526003
MFLX07147-30EA 188 USD
MFLX07147-30 MFLX07147-50 MFLX07147-40
Economical Submersible Pumps, Low-Flow Centrifugal
Pumps Submersible Pumps and Sump Pumps
Versatile—operate pumps in either a vertical or horizontal position.
  • Compact and economical
  • Screened inlet helps prevent clogging
Low-flow centrifugal submersible pumps for mild chemicals such as mild acids, alkalis, and hard water solutions compatible with pump materials. Use for water transfer or recirculation, water display, air conditioners, and machine tool coolants up to 100 SSU. Oil-filled for longer motor life.
Note: Pumps must be fully submersed when operating and not for use with salt water or flammable solutions.
Delivery information: Includes 12' power cord; 115 VAC models include grounded plug.