Ryton® PPS Sealless Chemical-Resistance Centrifugal Pumps
580603 584604 582603 587040 586604 585604 582614 583603 582604 581603
MFLX07085-05EA 2238 USD
MFLX07085-05 MFLX07085-18 MFLX07085-28 MFLX07085-08 MFLX07085-40 MFLX07085-12 MFLX07085-45 MFLX07085-22 MFLX07085-25 MFLX07085-02
Ryton® PPS Sealless Chemical-Resistance Centrifugal Pumps
Pumps Centrifugal Pumps
Ryton PPS construction is excellent with highly corrosive chemicals and solutions.
  • Leak-proof, sealless magnetic drive
  • Carbon bushing has run dry capability up to 8 hours—depending on fluid
  • Pump heads can be easily rotated, cleaned, or serviced without any special tools needed
These Ryton® PPS magnetic drive pumps ideal for pumping highly corrosive chemicals and solutions. Pumps are constructed of Ryton PPS body and impeller, ceramic shaft, Viton® O-rings, and carbon bushings.
Flow rates at 50 Hz will be 5/6 of flow rates shown.