4-Methylcyclohexene-1-ylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methyl-1-cyclohexen-1-ylboronic acid

43143-0010
TS43143-0010EA 319.01 USD
TS43143-0010
4-Methylcyclohexene-1-ylboronic acid 97%
4-Methylcyclohexene-1-ylboronic acid
Formula: C₇H₁₃BO₂
MW: 139.99
Melting Pt: 155…159 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659857
CAS Number: 850567-92-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR