4-Methyl-1-naphthaleneboronic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

43753-0100
TS43753-0100EA 249.73 USD
TS43753-0100
4-Methyl-1-naphthaleneboronic acid 96%
4-Methyl-1-naphthaleneboronic acid
Formula: C₁₁H₁₁BO₂
MW: 186.02
Melting Pt: 224…228 °C
Storage Temperature: Ambient
MDL Number: MFCD01632204
CAS Number: 103986-53-4

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR