6-Methoxy-2-naphthaleneboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Methoxy-2-naphthylboronic acid6-Methoxy-2-naphthylboronic Acid

39986-0050
TS39986-0050EA 93 USD
TS39986-0050
6-Methoxy-2-naphthaleneboronic acid 97%
6-Methoxy-2-naphthaleneboronic acid
Formula: C₁₁H₁₁BO₃
MW: 202.02 g/mol
Melting Pt: >295 °C
Storage Temperature: Ambient
MDL Number: MFCD03093087
CAS Number: 156641-98-4

Order Now

Specification Test Results

Appearance White to off-white to light brown Powder or crystalline powder
Infrared spectrum Authentic
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR