(±)-3-(Boc-amino)piperidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Aminopiperidine, 3-BOC protectedtert-Butyl piperidin-3-ylcarbamate

43751-0010 43751-0050
TS43751-0010EA 51.58 USD
TS43751-0010 TS43751-0050
(±)-3-(Boc-amino)piperidine 97%
(±)-3-(Boc-amino)piperidine
Formula: C₁₀H₂₀N₂O₂
MW: 200.28
Melting Pt: 104.6…112.6 °C
Storage Temperature: Ambient
MDL Number: MFCD03839941
CAS Number: 172603-05-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR