N-Boc-N-methylethylenediamine ≥97%

Danger

Synonyms: N-Boc-(2-Aminoethyl)-N-methylamineN-Methylethane-1,2-diamine, N-BOC protected

43734-0050
TS43734-0050EA 916 USD
TS43734-0050
N-Boc-N-methylethylenediamine ≥97%
N-Boc-N-methyl-ethylenediamine
Formula: C₈H₁₈N₂O₂
MW: 174.24 g/mol
Density: 0.975 g/cm³
Flash Pt: >100 °C
Storage Temperature: Ambient
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR