1-Cyclopentene-1-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

39683-0010 39683-0050
TS39683-0010EA 76.44 USD
TS39683-0010 TS39683-0050
1-Cyclopentene-1-carboxylic acid 98%
1-Cyclopentene-1-carboxylic acid

Order Now

Specification Test Results

Appearance White to yellow to orange Powder
Infrared spectrum Conforms
Melting point 118 to 122 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR