(2S)-(+)-2,3-Epoxypropyl toluene-4-sulphonate 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (2S)-(+)-Glycidyl tosylate(2S)-(+)-Glycidyl p-toluenesulphonate

26458-0250 26458-0050
TS26458-0250EA 420.96 USD
TS26458-0250 TS26458-0050
(2S)-(+)-2,3-Epoxypropyl toluene-4-sulphonate 99+%
(2S)-(+)-2,3-Epoxypropyl toluene-4-sulphonate
Formula: C₁₀H₁₂O₄S
MW: 228.26
Melting Pt: 45…48 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064489
CAS Number: 70987-78-9
EINECS: 417-210-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 45 to 48 °C
GC ≥99.0 %
Specific optical rotation +16° to +18° (20 °C, 589 nm) (c=2.5, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR