Methyl-4-bromocrotonate 85% tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
12645-1000
12645-5000
12645-0250
TS12645-5000EA
1250.63
USD
TS12645-5000
TS12645-0250
TS12645-1000
Methyl-4-bromocrotonate 85% tech.
Methyl 4-bromocrotonate
Formula:
C₅H₇BrO₂ MW: 179.01 Boiling Pt: 83…85 °C (13 mmHg) Density: 1.52 g/cm³ Flash Pt: 91 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000246 EINECS: 214-251-0 UN: 1760 ADR: 8,III |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4970 to 1.5050 (20°C, 589 nm) |
GC | ≥80 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...