Diethyl isopropylidenemalonate 97%

44331-0050
TS44331-0050EA 38.01 USD
TS44331-0050
Diethyl isopropylidenemalonate 97%
Diethyl isopropylidenemalonate
Formula: C₁₀H₁₆O₄
Boiling Pt: 175…178 °C (120 mmHg)
Density: 1.021 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00009147
CAS Number: 6802-75-1

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR