1,3-Phenylenediacetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

13050-0010 13050-0050 13050-0250
TS13050-0050EA 132.48 USD
TS13050-0050 TS13050-0250 TS13050-0010
1,3-Phenylenediacetic acid 97%
1,3-Phenylenediacetic acid
Formula: C₁₀H₁₀O₄
Melting Pt: 173…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00004341
CAS Number: 19806-17-8
EINECS: 243-332-3

Order Now

Specification Test Results

Appearance White to yellow Powder or crystals
Infrared spectrum Conforms
Melting point 171 to 179 °C
Titration with NaOH 96.0 to 104.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR