Ethyl vinyl ether 99% stabilized

Supplier: Thermo Scientific Chemicals
Danger

11908-0025 11908-2500 11908-0010
TS11908-0025EA 178 USD
TS11908-0025 TS11908-2500 TS11908-0010
Ethyl vinyl ether 99% stabilized
Ethyl vinyl ether
Formula: C₄H₈O
Boiling Pt: 36 °C (760 mmHg)
Melting Pt: –116 °C
Density: 0.75 g/cm³
Flash Pt: –45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009248
CAS Number: 109-92-2
EINECS: 203-718-4
UN: 1302
ADR: 3,I

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3760 to 1.3780 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer 0.1 % KOH

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR