4-Formylmorpholine 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Morpholine-4-carbaldehydeN-Formylmorpholine

13380-5000 13380-1000 13380-0050
TS13380-1000EA 45 USD
TS13380-1000 TS13380-5000 TS13380-0050
4-Formylmorpholine 99+%
4-Formylmorpholine
Formula: C₅H₉NO₂
MW: 115.13 g/mol
Melting Pt: 23 °C
Density: 1.14 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00006170
CAS Number: 4394-85-8
EINECS: 224-518-3

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4855 to 1.4875 (20°C, 589 nm)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR