2,3-Dimethylnaphthalene 97%

Supplier: Thermo Scientific Chemicals

11645-0010 11645-0050
TS11645-0010EA 56 USD
TS11645-0010 TS11645-0050
2,3-Dimethylnaphthalene 97%
2,3-Dimethylnaphthalene
Formula: C₁₂H₁₂
MW: 156.23 g/mol
Boiling Pt: 269 °C (760 mmHg)
Melting Pt: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00004119
CAS Number: 581-40-8

Order Now

Specification Test Results

Appearance Off-white to cream Crystalline powder
Infrared spectrum Conforms
Melting point 102 to 106 °C
GC ≥96.0 %
Nitrate (NO3) ≤0.003 %
Sulfate (SO4) ≤0.01 %
Calcium (Ca) ≤0.001 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.0005 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR