1-Cyclohexene-1-carboxylic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

29175-0025
TS29175-0025EA 104.98 USD
TS29175-0025
1-Cyclohexene-1-carboxylic acid 96%
1-Cyclohexene-1-carboxylic acid
Formula: C₇H₁₀O₂
Boiling Pt: 137 °C (15 mmHg)
Melting Pt: 35…39 °C
Storage Temperature: Ambient
MDL Number: MFCD00001545
CAS Number: 636-82-8

Order Now

Specification Test Results

Appearance White to light yellow Crystalline mass or liquid
Infrared spectrum Conforms
HPLC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR