Phenylbutazone 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Butyl-1,2-diphenylpyrazolidine-3,5-dione

19931-0050
TS19931-0050EA 38.98 USD
TS19931-0050
Phenylbutazone 99+%
Phenylbutazone
Formula: C₁₉H₂₀N₂O₂
MW: 308.38 g/mol
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00005500
CAS Number: 50-33-9
EINECS: 200-029-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 104 to 107 °C
Titration with NaOH ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR