Sodium 3-nitrobenzenesulfonate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium 3-nitrobenzenesulphonate3-Nitrobenzenesulfonic acid, sodium salt3-Nitrobenzenesulphonic acid, sodium salt

20912-0025 20912-0010
TS20912-0025EA 158 USD
TS20912-0025 TS20912-0010
Sodium 3-nitrobenzenesulfonate 99%
Sodium 3-nitrobenzenesulphonate
Formula: C₆H₄NNaO₅S
MW: 225.16 g/mol
Melting Pt: 350 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007490
CAS Number: 127-68-4
EINECS: 204-857-3

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
pH 6 to 10 (1 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR