1,1'-Binaphthyl 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,1'-Binaphthalene

40224-0050
TS40224-0050EA 126 USD
TS40224-0050
1,1'-Binaphthyl 97%
1,1'-Binaphthyl
Formula: C₂₀H₁₄
MW: 254.33 g/mol
Boiling Pt: 405 °C (1013 hPa)
Melting Pt: 143…146 °C
MDL Number: MFCD00041740
CAS Number: 604-53-5
EINECS: 210-070-6

Order Now

Specification Test Results

Appearance White to brown Powder
Infrared spectrum Conforms
GC ≥96.0 %
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR