Cyclododecanol 98+%

Supplier: Thermo Scientific Chemicals

11096-5000 11096-1000
TS11096-1000EA 42.92 USD
TS11096-1000 TS11096-5000
Cyclododecanol 98+%
Cyclododecanol
Formula: C₁₂H₂₄O
MW: 184.32 g/mol
Boiling Pt: 272.7 °C (1013 hPa)
Melting Pt: 76…79 °C
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00003717
CAS Number: 1724-39-6
EINECS: 217-031-2
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 76 to 80 °C
GC ≥98.0 %
Color scale ≤15 APHA
Titration with HCl ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR