1-Hexanethiol 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: n-Hexanethioln-Hexyl mercaptan1-Mercaptohexane

21527-0050 21527-1000 21527-5000
TS21527-1000EA 99 USD
TS21527-1000 TS21527-5000 TS21527-0050
1-Hexanethiol 96%
1-Hexanethiol
Formula: C₆H₁₄S
MW: 118.24 g/mol
Boiling Pt: 150…154 °C (760 mmHg)
Melting Pt: 81…80 °C
Density: 0.84 g/cm³
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00004909
CAS Number: 111-31-9
EINECS: 203-857-0
UN: 3071
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4480 to 1.4520 (20°C, 589 nm)
Density (20°C) 0.836 to 0.844 g/ml
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR