4-((tert-Butoxycarbonyl)amino)benzoic acid 97%

Warning

Synonyms: 4-(Boc-amino)benzoic AcidBoc-4-Abz-OH

42618-0050 42618-0250
TS42618-0050EA 102.88 USD
TS42618-0050 TS42618-0250
4-((tert-Butoxycarbonyl)amino)benzoic acid 97%
4-((tert-Butoxycarbonyl)amino)benzoic acid
Formula: C₁₂H₁₅NO₄
MW: 237.3 g/mol
Melting Pt: 179…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00037428
CAS Number: 66493-39-8

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR