1-Boc-piperidine-3-carboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-BOC-3-Piperidinecarboxylic acid
, Boc-Nip-OH
, 1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid
, 1-[(tert-butoxy)carbonyl]piperidine-3-carboxylic acid
, N-Boc-DL-nipecotic acid
39958-0250
39958-0050
TS39958-0250EA
135
USD
TS39958-0250
TS39958-0050
1-Boc-piperidine-3-carboxylic acid 98%
1-Boc-piperidine-3-carboxylic acid
Formula:
C₁₁H₁₉NO₄ MW: 229.3 g/mol Melting Pt: 159…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00673775 CAS Number: 84358-12-3 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 159 to 162 °C |
Titration with NaOH | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...