L-Methionine sulfone 98%

Supplier: Thermo Scientific Chemicals

43900-0010 43900-0050
TS43900-0010EA 74.74 USD
TS43900-0010 TS43900-0050
L-Methionine sulfone 98%
L-Methionine sulphone
Formula: C₅H₁₁NO₄S
Melting Pt: 242.3…243.1 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00066020
CAS Number: 7314-32-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
TLC ≥97.5 %
Elemental Analysis 32.31 to 33.97 % C
Elemental Analysis 7.54 to 7.92 % N
Specific optical rotation +11.7° to +13.7° (24°C, 589 nm) (c=2, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR