Okadaic acid sodium salt 98%, high purity

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 9,10-deepithio-9,10-didehydroacanthifolicin sodium saltOA sodium salt

32840-0001
TS32840-0001EA 412.78 USD
TS32840-0001
Okadaic acid sodium salt 98%, high purity
Okadaic acid sodium salt
Formula: C₄₄H₆₇NaO₁₃
MW: 827.00 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00210208
CAS Number: 209266-80-8
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear white to off-white Solid or film
TLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR