Retinoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid
, all-trans-Retinoic acid
, Vitamin A acid
, ATRA
, Tretinoin
20734-0050
20734-1000
20734-0010
TS20734-0050EA
683
USD
TS20734-0050
TS20734-1000
TS20734-0010
Retinoic acid 97%
Retinoic acid
Formula:
C₂₀H₂₈O₂ MW: 300.44 g/mol Melting Pt: 179…184 °C Storage Temperature: Freezer |
MDL Number:
MFCD00001551 CAS Number: 302-79-4 EINECS: 206-129-0 |
Specification Test Results
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 179 to 184 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...