Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) hydrate 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 5-CTP-Na2CTP

22622-5000
TS22622-5000EA 404.51 USD
TS22622-5000
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) hydrate 95%
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) hydrate
Formula: C₉H₁₄N₃Na₂O₁₄P₃·xH₂O
Melting Pt: 215…218 °C
Storage Temperature: Freezer
MDL Number: MFCD00150809
CAS Number: 652154-13-7
EINECS: 252-849-3

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤10 % (105°C, 5 h)
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR