Adenosine 99+%
Supplier: Thermo Scientific Chemicals
Synonyms:
Adenine riboside
, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol
, Adenine-9-β-D-ribofuranoside
, 9-β-D-Ribofuranosyladenine
16404-5000
16404-0250
16404-1000
16404-0050
TS16404-5000EA
1121
USD
TS16404-5000
TS16404-0250
TS16404-0050
TS16404-1000
Adenosine 99+%
Adenosine
Formula:
C₁₀H₁₃N₅O₄ MW: 267.24 g/mol Melting Pt: 233…238 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005752 CAS Number: 58-61-7 EINECS: 200-389-9 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 233 to 238 °C |
Loss on drying | ≤0.5 % (105 °C, 2 hrs) |
Sulfated ash | ≤0.1 % |
HPLC | ≥99.0 % |
UV spectrophotometry | 98.0 to 102.0 % (lambda max. 259 nm, pH 7) |
UV spectrophotometry | molar absorptivity 15400 +/- 2% , on dry basis |
UV transmittance | ≥98 % (c=0.5, H2O, 430 nm) 1 cm cell |
Ratio of Absorbances | 0.76 to 0.80 (A250/A260) |
Ratio of Absorbances | 0.14 to 0.16 (A280/A260) |
Nitrogen (N) | 25.69 to 26.73 % (on dry basis) |
Heavy metals (as Pb) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...