CAPSO sodium salt (3-(cyclohexylamino)-2-hydroxypropanesulfonic acid sodium salt) 98%

Supplier: Thermo Scientific Chemicals
Danger

44413-1000
TS44413-1000EA 132.08 USD
TS44413-1000
CAPSO sodium salt (3-(cyclohexylamino)-2-hydroxypropanesulfonic acid sodium salt) 98%
CAPSO sodium salt (3-(Cyclohexylamino)-2-hydroxypropanesulphonic acid sodium salt)
Formula: C₉H₁₈NNaO₄S
Melting Pt: 206…211 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00070063
CAS Number: 102601-34-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤5.0 %
Solubility (10 % in water) Clear colorless
UV (0.1 M in water)
UV at 260 nm A: ≤0.050
UV at 280 nm A: ≤0.050
pH 10.9 to 11.6 (1 % in water at 25°C)
Titration with HClO4 97.5 to 102.5 % (on dry basis)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR